2-Chloro-3-formylpyridine 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Chloropyridine-3-carboxaldehyde, 2-Chloronicotinaldehyde, 2-Chloro-3-pyridinecarboxaldehyde

42979-0010 42979-0050
TS42979-0010EA 35 USD
TS42979-0010 TS42979-0050
2-Chloro-3-formylpyridine 98%
2-Chloro-3-formylpyridine
Formula: C₆H₄ClNO
MW: 141.56 g/mol
Melting Pt: 47…50 °C
Storage Temperature: Ambient
MDL Number: MFCD01315308
CAS Number: 36404-88-3
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Light yellow Crystalline powder or solid
Infrared spectrum Conforms
Melting point 47 to 52 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR