6-Methoxypyridine-3-sulfonyl chloride 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 6-Methoxy-pyridine-3-sulphonyl chloride, 3-Pyridinesulphonylchloride, 6-methoxy-, 3-(Chlorosulphonyl)-6-methoxypyridine

42941-0010 42941-0050
TS42941-0010EA 239 USD
TS42941-0010 TS42941-0050
6-Methoxypyridine-3-sulfonyl chloride 97%
6-Methoxypyridine-3-sulfonyl chloride
Formula: C₆H₆ClNO₃S
MW: 207.628 g/mol
Melting Pt: 42…45 °C
Storage Temperature: Freezer
MDL Number: MFCD06739108
CAS Number: 312300-42-8
UN: 3261
ADR: 8,III

Order Now

Specification Test Results

Appearance White to off-white Low melting crystalline solid
Infrared spectrum Conforms
Melting point 42 to 46 °C
GC ≥96.0 %
Absorbance ≤0.01 (water, 1 cm, 254 nm)
Chloride (Cl) ≤0.1 ppm
Nitrate (NO3) ≤0.2 ppm
Phosphate (PO4) ≤0.05 ppm
Sulfate (SO4) ≤1 ppm
Silicate (as SiO2) ≤0.01 ppm
Aluminium (Al) ≤0.02 ppm
Ammonium (NH4) ≤0.01 ppm
Antimony (Sb) ≤0.01 ppm
Barium (Ba) ≤0.01 ppm
Bismuth (Bi) ≤0.01 ppm
Boron (B) ≤0.01 ppm
Cadmium (Cd) ≤0.01 ppm
Calcium (Ca) ≤0.1 ppm
Chromium (Cr) ≤0.01 ppm
Cobalt (Co) ≤0.01 ppm
Copper (Cu) ≤0.01 ppm
Gold (Au) ≤0.01 ppm
Iron (Fe) ≤0.01 ppm
Lithium (Li) ≤0.02 ppm
Magnesium (Mg) ≤0.05 ppm
Manganese (Mn) ≤0.01 ppm
Mercury (Hg) ≤0.05 ppm
Molybdenum (Mo) ≤0.01 ppm
Nickel (Ni) ≤0.01 ppm
Potassium (K) ≤0.05 ppm
Selenium (Se) ≤0.01 ppm
Silver (Ag) ≤0.01 ppm
Sodium (Na) ≤0.1 ppm
Strontium (Sr) ≤0.05 ppm
Tin (Sn) ≤0.01 ppm
Titanium (Ti) ≤0.01 ppm
Vanadium (V) ≤0.01 ppm
Zinc (Zn) ≤0.05 ppm
Zirconium (Zr) ≤0.01 ppm
Heavy metals (as Pb) ≤0.01 ppm
KMnO4-reducing substance Passes test
Conductivity ≤6 µS/cm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR