ω-Bromo-3-hydroxyacetophenone 96%
Supplier: Thermo Scientific Chemicals
|
Danger
|
42862-0010
42862-0050
TS42862-0010EA
102
USD
TS42862-0010
TS42862-0050
ω-Bromo-3-hydroxyacetophenone 96%
ω-Bromo-3-hydroxyacetophenone
Formula:
C₈H₇BrO₂ Melting Pt: 70…74 °C Storage Temperature: Ambient |
MDL Number:
MFCD05664326 CAS Number: 2491-37-4 UN: 3261 ADR: 8,II |
Specification Test Results
Appearance | Brown Powder |
Infrared spectrum | Conforms |
HPLC | ≥95 % |
NMR (1H-NMR) | Consistent with structure |
Specific optical rotation | -22° to -26° (20°C, 589 nm) (c=1, ethanol) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...