Methyl tricosanoate 99+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Tricosanoic acid methyl ester

42845-0010 42845-0050
TS42845-0010EA 418 USD
TS42845-0010 TS42845-0050
Methyl tricosanoate 99+%
Methyl tricosanoate
Formula: C₂₄H₄₈O₂
Melting Pt: 53 °C
Flash Pt: >113 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00009349
CAS Number: 2433-97-8

Order Now

Specification Test Results

Appearance White Powder
Infrared spectrum Conforms
Melting point 53 to 56 °C
GC ≥99.0 %
NMR (1H-NMR) Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR