ω-Bromo-2-chloroacetophenone 95%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
α-Bromo-2-chloroacetophenone, Bromomethyl o-chlorophenyl ketone, 2-Bromo-1-(2-chlorophenyl)ethanone, 2-Bromo-2'-chloroacetophenone, 2-Chlorophenacyl bromide
42789-0010
42789-0050
TS42789-0010EA
69
USD
TS42789-0010
TS42789-0050
ω-Bromo-2-chloroacetophenone 95%
ω-Bromo-2-chloroacetophenone
Formula:
C₈H₆BrClO Boiling Pt: 162…166 °C (20 mmHg) Flash Pt: >110 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00832993 CAS Number: 5000-66-8 UN: 3265 ADR: 8,II |
Specification Test Results
Appearance | Colorless to light yellow Liquid |
Infrared spectrum | Conforms |
GC | ≥94 % |
NMR (1H-NMR) | Consistent with structure |
Chloride (Cl) | ≤0.001 % |
Oxalate (C2O4) | (limit about 0.05%) Passes test |
Phosphate (PO4) | ≤0.001 % |
Sulfate (SO4) | ≤0.002 % |
Iron (Fe) | ≤3 ppm |
Lead (Pb) | ≤2 ppm |
Insoluble matter | ≤0.005 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...