4-Bromo-1-indanone 97%

Supplier: Thermo Scientific Chemicals
Warning

42770-0010 42770-0050
TS42770-0010EA 50 USD
TS42770-0010 TS42770-0050
4-Bromo-1-indanone 97%
4-Bromo-1-indanone
Formula: C₉H₇BrO
Melting Pt: 95…99 °C
Storage Temperature: Ambient
MDL Number: MFCD01719772
CAS Number: 15115-60-3

Order Now

Specification Test Results

Appearance Orange-brown Crystalline powder
Infrared spectrum Conforms
Melting point 94 to 98 °C
GC ≥96.0 %
NMR (1H-NMR) Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR