2-Chloro-4-methoxypyrimidine 98%

Supplier: Thermo Scientific Chemicals
Warning

42682-0010 42682-0050
TS42682-0010EA 45 USD
TS42682-0010 TS42682-0050
2-Chloro-4-methoxypyrimidine 98%
2-Chloro-4-methoxypyrimidine
Formula: C₅H₅ClN₂O
MW: 144.56 g/mol
Melting Pt: 54…57 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00194055
CAS Number: 22536-63-6

Order Now

Specification Test Results

Appearance White to yellow Crystalline powder
Infrared spectrum Conforms
Melting point 54 to 57 °C
GC ≥97.5 %
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR