3,3-Diethoxy-1-propanol 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3,3-diethoxypropan-1-ol, 3-hydroxypropionaldehyde diethyl acetal, 1-Propanol,3,3-diethoxy-

42672-0010 42672-0050
TS42672-0010EA 55 USD
TS42672-0010 TS42672-0050
3,3-Diethoxy-1-propanol 98%
3,3-Diethoxy-1-propanol
Formula: C₇H₁₆O₃
MW: 148.202 g/mol
Boiling Pt: 90…93 °C (14 mmHg)
Density: 0.941 g/cm³
Flash Pt: 91 °C
Storage Temperature: Ambient
MDL Number: MFCD00074850
CAS Number: 16777-87-0

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR