Methyl undecanoate 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Undecanoic acid methyl ester, Undecylic acid methyl ester

42658-0050 42658-0250
TS42658-0050EA 55 USD
TS42658-0050 TS42658-0250
Methyl undecanoate 97%
Methyl undecanoate
Formula: C₁₂H₂₄O₂
MW: 200.32 g/mol
Boiling Pt: 246…248 °C (760 mmHg)
Density: 0.872 g/cm³
Flash Pt: 109 °C
Storage Temperature: Ambient
MDL Number: MFCD00008957
CAS Number: 1731-86-8

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
GC ≥96.0 %
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR