3-Chloro-4-formylpyridine 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Chloroisonicotinaldehyde, 3-Chloro-4-pyridinecarboxaldehyde, 3-Chloropyridine-4-carboxaldehyde

42655-0010 42655-0050
TS42655-0010EA 49 USD
TS42655-0010 TS42655-0050
3-Chloro-4-formylpyridine 97%
3-Chloro-4-formylpyridine
Formula: C₆H₄ClNO
MW: 141.56 g/mol
Boiling Pt: 225 °C
Melting Pt: 67…68 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06200712
CAS Number: 72990-37-5

Order Now

Specification Test Results

Appearance White to yellow Crystalline powder
Infrared spectrum Conforms
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR