Methyl-4-iodobutanoate 95% stabilized

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Methyl 4-iodobutanoate, Methyl 4-iodobutyrate

42637-0010 42637-0050
TS42637-0010EA 98 USD
TS42637-0010 TS42637-0050
Methyl-4-iodobutanoate 95% stabilized
Methyl-4-iodobutanoate
Formula: C₅H₉IO₂
MW: 228.029 g/mol
Boiling Pt: 80…83 °C (11 mmHg)
Density: 1.689 g/cm³
Flash Pt: 99 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00674066
CAS Number: 14273-85-9

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
GC ≥94 %
Stabilizer Copper
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR