Ethyl glyoxylate ∼50% in toluene, Technical Grade
Supplier: Thermo Scientific Chemicals
|
Danger
|
42576-0010
42576-5000
TS42576-0010EA
309
USD
TS42576-0010
TS42576-5000
Ethyl glyoxylate ∼50% in toluene, Technical Grade
Ethyl glyoxylate
Formula:
C₄H₆O₃ MW: 102.09 g/mol Boiling Pt: 110 °C (760 mmHg) Density: 1.032 g/cm³ Flash Pt: 7 °C Storage Temperature: Ambient |
MDL Number:
MFCD00044009 CAS Number: 924-44-7 UN: 1294 ADR: 3,II |
Specification Test Results
Appearance | Clear colorless to light yellow Solution |
Titration excess HCl | ≥48 % (Aldehyde function) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...