2-Amino-4'-methoxyacetophenone hydrochloride 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
42571-0010
42571-0050
TS42571-0010EA
52
USD
TS42571-0010
TS42571-0050
2-Amino-4'-methoxyacetophenone hydrochloride 97%
2-Amino-4'-methoxyacetophenone hydrochloride
Formula:
C₉H₁₂ClNO₂ Melting Pt: 190…193 °C Storage Temperature: Ambient |
MDL Number:
MFCD00193078 CAS Number: 3883-94-1 |
Specification Test Results
Appearance | White to yellow to tan Crystalline powder |
Infrared spectrum | Conforms |
Titration Argentometric | ≥96.0 % |
HPLC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...