2-Methyl-3-buten-1-ol 98%

Supplier: Thermo Scientific Chemicals
Warning

42569-0010 42569-0050
TS42569-0010EA 100 USD
TS42569-0010 TS42569-0050
2-Methyl-3-buten-1-ol 98%
2-Methyl-3-buten-1-ol
Formula: C₅H₁₀O
MW: 86.134 g/mol
Boiling Pt: 120…121 °C (760 mmHg)
Density: 0.835 g/cm³
Flash Pt: 33 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00009985
CAS Number: 4516-90-9
UN: 1987
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4240 to 1.4300 (20 °C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR