Methyl-4-acetylbenzoate 99+%

Supplier: Thermo Scientific Chemicals

Synonyms: Methyl 4-acetylbenzoate

42477-0010 42477-0050
TS42477-0010EA 39 USD
TS42477-0010 TS42477-0050
Methyl-4-acetylbenzoate 99+%
Methyl 4-acetylbenzoate
Formula: C₁₀H₁₀O₃
Melting Pt: 93…95 °C
Storage Temperature: Ambient
MDL Number: MFCD00216474
CAS Number: 3609-53-8

Order Now

Specification Test Results

Appearance White to light yellow Powder
Infrared spectrum Conforms
Melting point 93 to 95 °C
GC ≥99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR