2,3,4-Trihydroxybenzaldehyde 98%

Supplier: Thermo Scientific Chemicals
Warning

42179-0050 42179-0250 42179-1000
TS42179-0050EA 53 USD
TS42179-0050 TS42179-0250 TS42179-1000
2,3,4-Trihydroxybenzaldehyde 98%
2,3,4-Trihydroxybenzaldehyde
Formula: C₇H₆O₄
Melting Pt: 159…162 °C
Storage Temperature: Ambient
MDL Number: MFCD00003325
CAS Number: 2144-08-3
EINECS: 218-404-2

Order Now

Specification Test Results

Appearance Beige to light brown Powder
Infrared spectrum Conforms
Melting point 159 to 162 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR