Tetraethylene glycol monomethyl ether 98%

Supplier: Thermo Scientific Chemicals

Synonyms: mPEG4-OH, 2,5,8,11-Tetraoxatridecan-13-ol

42035-0050 42035-0250
TS42035-0050EA 80 USD
TS42035-0050 TS42035-0250
Tetraethylene glycol monomethyl ether 98%
Tetraethylene glycol monomethyl ether
Formula: C₉H₂₀O₅
MW: 208.25 g/mol
Boiling Pt: 158…160 °C (5 mmHg)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00041756
CAS Number: 23783-42-8
EINECS: 245-883-5

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4440 to 1.4460 (20 °C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR