Paraffin wax, granular, pure
Supplier: Thermo Scientific Chemicals
Synonyms:
Wax Paraffin, Paraffin wax
41677-0020
41677-0100
TS41677-0020EA
119
USD
TS41677-0020
TS41677-0100
Paraffin wax, granular, pure
Paraffin
Boiling Pt:
322 °C (760 mmHg) Melting Pt: 58…62 °C Flash Pt: 198 °C Storage Temperature: Ambient |
CAS Number:
8002-74-2 |
Specification Test Results
Appearance | White Pellets |
Infrared spectrum | Conforms |
Congealing point | 58 °C to 62 °C |
Titration Argentometric | ≥98.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...