Sodium palmitate 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
Palmitic Acid Sodium Salt
41670-0050
41670-0250
41670-1000
TS41670-0050EA
75
USD
TS41670-0050
TS41670-0250
TS41670-1000
Sodium palmitate 98%
Sodium palmitate
Formula:
C₁₆H₃₂O₂ MW: 278.41 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00002749 CAS Number: 408-35-5 |
Specification Test Results
Appearance | White Powder |
Infrared spectrum | Conforms |
Loss on drying | ≤2 % (110°C, 4 hrs) |
Titration with HClO4 | 97.5 to 102.5 % (On dry substance) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...