4-Nitrophthalonitrile 99%

Supplier: Thermo Scientific Chemicals
Warning

41615-0050 41615-0250
TS41615-0050EA 205 USD
TS41615-0050 TS41615-0250
4-Nitrophthalonitrile 99%
4-Nitrophthalonitrile
Formula: C₈H₃N₃O₂
Melting Pt: 140…144 °C
Storage Temperature: Ambient
MDL Number: MFCD00040301
CAS Number: 31643-49-9
EINECS: 250-748-9

Order Now

Specification Test Results

Appearance Light yellow to beige to light green to light grey Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 140 to 144 °C
HPLC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR