Methyl palmitate 95%

Supplier: Thermo Scientific Chemicals

Synonyms: Hexadecylic acid methyl ester, Methyl hexadecanoate, Palmitic acid methyl ester, Hexadecanoic acid methyl ester, Palmitic Acid Methyl Ester

41468-1000 41468-5000
TS41468-1000EA 28 USD
TS41468-1000 TS41468-5000
Methyl palmitate 95%
Methyl palmitate
Formula: C₁₇H₃₄O₂
MW: 270.46 g/mol
Boiling Pt: 417 °C (1013 hPa)
Melting Pt: 28 °C
Flash Pt: >110 °C
MDL Number: MFCD00008994
CAS Number: 112-39-0
EINECS: 203-966-3

Order Now

Specification Test Results

Appearance Clear colorless Liquid or low melting solid
Infrared spectrum Conforms
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR