Methyl oleate, technical

Supplier: Thermo Scientific Chemicals

Synonyms: cis-Oleic Acid Methyl Ester, Oleic acid methyl ester

41464-0010 41464-0050 41464-5000
TS41464-0010EA 58 USD
TS41464-0010 TS41464-0050 TS41464-5000
Methyl oleate, technical
Methyl oleate
Formula: C₁₉H₃₆O₂
MW: 296.49 g/mol
Boiling Pt: 186 °C (9 mmHg)
Melting Pt: –20 °C
Density: 0.87 g/cm³
Flash Pt: 170 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00009578
CAS Number: 112-62-9
EINECS: 203-992-5
Merck Index: 12,06965

Order Now

Specification Test Results

Appearance Clear yellow to amber Viscous liquid
Infrared spectrum Conforms
GC ≥55 %
HPLC ≥98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR