Methyl octanoate 99%

Supplier: Thermo Scientific Chemicals

Synonyms: Caprylic acid methyl ester, Octanoic acid methyl ester, Methyl caprylate

41462-0250 41462-1000 41462-5000
TS41462-0250EA 53 USD
TS41462-0250 TS41462-1000 TS41462-5000
Methyl octanoate 99%
Methyl octanoate
Formula: C₉H₁₈O₂
MW: 158.24 g/mol
Boiling Pt: 79 °C (12 mmHg)
Melting Pt: –36.7 °C
Density: 0.878 g/cm³
Flash Pt: 73 °C
MDL Number: MFCD00009551
CAS Number: 111-11-5
EINECS: 203-835-0

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4165 to 1.4185 (20 °C, 589 nm)
GC ≥98.5 %
Specific gravity (20°C) 0.868 to 0.888
Acid value ≤1 mg KOH/g

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR