Methyl malonyl chloride 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Methyl 3-chloro-3-oxopropanoate
41451-0050
41451-0250
41451-1000
TS41451-0050EA
65
USD
TS41451-0050
TS41451-0250
TS41451-1000
Methyl malonyl chloride 97%
Methyl malonyl chloride
Formula:
C₄H₅ClO₃ Boiling Pt: 57…59 °C (12 mmHg) Flash Pt: 80 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00013657 CAS Number: 37517-81-0 EINECS: 253-540-6 UN: 3265 ADR: 8,II |
Specification Test Results
Appearance | Clear yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4310 to 1.4350 (20 °C, 589 nm) |
Titration RCOCl | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...