Methyl butyrate 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Butyric acid methyl ester, Butanoic acid methyl ester

41414-0050 41414-1000 41414-2500
TS41414-0050EA 32 USD
TS41414-0050 TS41414-1000 TS41414-2500
Methyl butyrate 99%
Methyl butyrate
Formula: C₅H₁₀O₂
MW: 102.13 g/mol
Boiling Pt: 102 °C
Melting Pt: –84 °C
Density: 0.897 g/cm³
Flash Pt: 12 °C
MDL Number: MFCD00009391
CAS Number: 623-42-7
EINECS: 210-792-1
UN: 1237
ADR: 3,II
Merck Index: 13,06062

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.3850 to 1.3900 (20 °C, 589 nm)
Density (20°C) 0.893 to 0.899 g/ml
GC ≥98.5 %
Acid value ≤1 mg KOH/g

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR