Sodium laurate 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Sodium Laurate, Lauric acid sodium salt, Sodium dodecanoate
41304-0050
41304-0250
41304-1000
TS41304-0050EA
50
USD
TS41304-0050
TS41304-0250
TS41304-1000
Sodium laurate 98%
Sodium laurate
Formula:
C₁₂H₂₃NaO₂ MW: 222.304 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00041754 CAS Number: 629-25-4 EINECS: 211-082-4 UN: 3077 ADR: 9 III |
Specification Test Results
Appearance | White to light yellow Powder or crystals |
Infrared spectrum | Conforms |
Water | ≤3 % |
GC | ≥97.5 % |
Sodium (Na) | 7.8 to 15.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...