4-Isopropylbenzoic acid 98+%

Supplier: Thermo Scientific Chemicals
Warning

41280-0050 41280-0250
TS41280-0050EA 61 USD
TS41280-0050 TS41280-0250
4-Isopropylbenzoic acid 98+%
4-Isopropylbenzoic acid
Formula: C₁₀H₁₂O₂
MW: 164.2 g/mol
Melting Pt: 116…120 °C
Storage Temperature: Ambient
MDL Number: MFCD00002564
CAS Number: 536-66-3
EINECS: 208-642-5
Merck Index: 12,02684

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 116 to 120 °C
HPLC ≥98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR