Acetoin dimer 96%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1,4-Dioxane-2,5-diol, 2,3,5,6-tetramethyl-

41195-1000 41195-5000
TS41195-1000EA 111 USD
TS41195-1000 TS41195-5000
Acetoin dimer 96%
Acetoin dimer
Formula: C₈H₁₆O₄
MW: 176.21 g/mol
Boiling Pt: 148 °C (1013 hPa)
Melting Pt: 90…91 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004521
CAS Number: 23147-57-1
EINECS: 208-174-1
UN: 1325
ADR: 4.1,III
Merck Index: 13,00065

Order Now

Specification Test Results

Appearance White to yellow Powder or crystals
Infrared spectrum Conforms
GC ≥95 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR