3,5-Heptanedione 99%

Supplier: Thermo Scientific Chemicals
Warning

41126-0050 41126-0250
TS41126-0050EA 78 USD
TS41126-0050 TS41126-0250
3,5-Heptanedione 99%
3,5-Heptanedione
Formula: C₇H₁₂O₂
Boiling Pt: 172…173 °C (711 mmHg)
Density: 0.945 g/cm³
Flash Pt: 57 °C
Storage Temperature: Ambient
MDL Number: MFCD00015186
CAS Number: 7424-54-6
EINECS: 231-054-5
UN: 1224
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4540 to 1.4570 (20 °C, 589 nm)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR