Glycolic acid 70%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-Hydroxy acid, 2-Hydroxyacetic acid

41103-0010 41103-0250 41103-2500
TS41103-0010EA 40 USD
TS41103-0010 TS41103-0250 TS41103-2500
Glycolic acid 70%
Glycolic acid
Formula: C₂H₄O₃
MW: 76.05 g/mol
Melting Pt: 10 °C
Density: 1.27 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00004312
CAS Number: 79-14-1
EINECS: 201-180-5
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear yellow to light amber Solution
Titration with NaOH 65 to 72 % w/w
GC ≥94 %
Enantiomeric excess ≥97.5 % (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR