Ethyl glycolate 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Ethyl Hydroxyacetate, Hydroxyacetic Acid Ethyl Ester, Glycolic Acid Ethyl Ester, Ethyl 2-hydroxyacetate
41008-0250
41008-1000
41008-5000
TS41008-0250EA
107
USD
TS41008-0250
TS41008-1000
TS41008-5000
Ethyl glycolate 95%
Ethyl glycolate
Formula:
C₄H₈O₃ Boiling Pt: 158…159 °C (760 mmHg) Density: 1.08 g/cm³ Flash Pt: 62 °C Storage Temperature: Ambient |
MDL Number:
MFCD00021970 CAS Number: 623-50-7 EINECS: 210-798-4 |
Specification Test Results
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4170 to 1.4200 (20 °C, 589 nm) |
GC | ≥94 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...