4'-Ethoxyacetophenone 99%

Supplier: Thermo Scientific Chemicals
Warning

40959-0050 40959-0250 40959-1000 40959-5000
TS40959-0050EA 34 USD
TS40959-0050 TS40959-0250 TS40959-1000 TS40959-5000
4'-Ethoxyacetophenone 99%
4'-Ethoxyacetophenone
Formula: C₁₀H₁₂O₂
MW: 164.2 g/mol
Boiling Pt: 268…269 °C (758 mmHg)
Melting Pt: 35…38 °C
Flash Pt: 134 °C
Storage Temperature: Ambient
MDL Number: MFCD00009095
CAS Number: 1676-63-7
EINECS: 216-825-6

Order Now

Specification Test Results

Appearance White to yellow Crystals and/or chunks or low melting solid
Infrared spectrum Conforms
Melting point 35 to 38 °C
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR