Dipropyleneglycol (mixture of isomers) 99%
Supplier: Thermo Scientific Chemicals
40915-0010
40915-0030
TS40915-0010EA
89
USD
TS40915-0010
TS40915-0030
Dipropyleneglycol (mixture of isomers) 99%
Dipropyleneglycol (mixture of isomers)
Formula:
C₆H₁₄O₃ Flash Pt: 138 °C Storage Temperature: Ambient |
MDL Number:
MFCD00051023 CAS Number: 25265-71-8 EINECS: 246-770-3 |
Specification Test Results
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4390 to 1.4420 (20 °C, 589 nm) |
GC | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...