1,4-Dinitrobenzene 98%

Supplier: Thermo Scientific Chemicals
Danger

40865-0050 40865-0250
TS40865-0050EA 57 USD
TS40865-0050 TS40865-0250
1,4-Dinitrobenzene 98%
1,4-Dinitrobenzene
Formula: C₆H₄N₂O₄
MW: 168.11 g/mol
Boiling Pt: 299
Melting Pt: 172
Density: 1.63 g/cm³
Flash Pt: 150 °C
Storage Temperature: Ambient
MDL Number: MFCD00007314
CAS Number: 100-25-4
EINECS: 202-833-7
UN: 3443
ADR: 6.1 II
Merck Index: 13,03301

Order Now

Specification Test Results

Appearance Ochre to orange Powder or crystals
Infrared spectrum Conforms
Melting point 171 to 175 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR