2,6-Dimethyl-3,5-heptanedione 99%

Supplier: Thermo Scientific Chemicals
Warning

40836-0050 40836-0250
TS40836-0050EA 130 USD
TS40836-0050 TS40836-0250
2,6-Dimethyl-3,5-heptanedione 99%
2,6-Dimethyl-3,5-heptanedione
Formula: C₉H₁₆O₂
MW: 156.22 g/mol
Boiling Pt: 66 °C (8 mmHg)
Density: 1.4586 g/cm³
Flash Pt: 60 °C
MDL Number: MFCD00015040
CAS Number: 18362-64-6
EINECS: 242-234-8
UN: 1224
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4560 to 1.4600 (20 °C, 589 nm)
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR