Diethyl oxalacetate sodium salt 95% pract.
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Sodium diethyl oxalacetate, Sodium Diethyl Oxalacetate, Oxalacetic Acid Diethyl Ester Sodium Salt
40737-1000
40737-5000
TS40737-1000EA
53
USD
TS40737-1000
TS40737-5000
Diethyl oxalacetate sodium salt 95% pract.
Diethyl oxalacetate sodium salt
Formula:
C₈H₁₂O₅ Storage Temperature: Ambient |
MDL Number:
MFCD00035571 CAS Number: 40876-98-0 |
Specification Test Results
Appearance | Beige to light orange Crystalline powder |
Infrared spectrum | Conforms |
Titration with HClO4 | ≥94 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...