Diethyl adipate 99%

Supplier: Thermo Scientific Chemicals

Synonyms: NSC 19160, Adipic acid diethyl ester, 1,6-Hexanedioic acid diethyl ester, NSC 3363

40717-1000 40717-5000
TS40717-1000EA 23 USD
TS40717-1000 TS40717-5000
Diethyl adipate 99%
Diethyl adipate
Formula: C₁₀H₁₈O₄
MW: 202.25 g/mol
Boiling Pt: 251
Melting Pt: -19.8
Density: 1 g/cm³
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00009215
CAS Number: 141-28-6
EINECS: 205-477-0
Merck Index: 13,00163

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4260 to 1.4280 (20 °C, 589 nm)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR