3,5-Dichlorosalicylaldehyde 98+%

Supplier: Thermo Scientific Chemicals
Warning

40700-0050 40700-0250
TS40700-0050EA 50 USD
TS40700-0050 TS40700-0250
3,5-Dichlorosalicylaldehyde 98+%
3,5-Dichlorosalicylaldehyde
Formula: C₇H₄Cl₂O₂
Melting Pt: 91…94 °C
Storage Temperature: Ambient
MDL Number: MFCD00003320
CAS Number: 90-60-8
EINECS: 202-005-5

Order Now

Specification Test Results

Appearance Light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 91 to 94 °C
Titration with TBAH ≥98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR