2,6-Dibromoquinone-4-chloroimide 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2,6-Dibromo-N-chloro-p-benzoquinoneimine

40647-0050 40647-0250
TS40647-0050EA 89 USD
TS40647-0050 TS40647-0250
2,6-Dibromoquinone-4-chloroimide 97%
2,6-Dibromoquinone-4-chloroimide
Formula: C₆H₂Br₂ClNO
Melting Pt: 76…83 °C
Density: 1.81 g/cm³
Storage Temperature: Refrigerator
MDL Number: MFCD00013792
CAS Number: 537-45-1
EINECS: 208-667-1
UN: 3224
ADR: 4.1,

Order Now

Specification Test Results

Appearance Yellow to ochre Powder or crystalline needles (May darken on storage)
Infrared spectrum Conforms
Melting point 76 to 83 °C
Titration Iodometric ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR