5-Chlorosalicylaldehyde 98%

Supplier: Thermo Scientific Chemicals
Warning

40503-0050 40503-0250 40503-1000
TS40503-0050EA 36 USD
TS40503-0050 TS40503-0250 TS40503-1000
5-Chlorosalicylaldehyde 98%
5-Chlorosalicylaldehyde
Formula: C₇H₅ClO₂
Boiling Pt: 105 °C (12 mmHg)
Melting Pt: 102…103 °C
Storage Temperature: Ambient
MDL Number: MFCD00003331
CAS Number: 635-93-8
EINECS: 211-244-4
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 99 to 103 °C
GC ≥97.5 %
Residue after evaporation ≤0.0005 %
Free amine ≤0.0002 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR