3-Bromo-1-propanol 92%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Bromopropan-1-ol, 1-Bromo-3-hydroxypropane

40320-0050 40320-0250
TS40320-0050EA 34 USD
TS40320-0050 TS40320-0250
3-Bromo-1-propanol 92%
3-Bromo-1-propanol
Formula: BrH₂CCH₂CH₂OH
MW: 138.99 g/mol
Boiling Pt: 62 °C (5 mmHg)
Density: 1.6 g/cm³
Flash Pt: 65 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00002942
CAS Number: 627-18-9
EINECS: 210-986-6

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4850 to 1.4900 (20 °C, 589 nm)
GC ≥90 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR