Desyl bromide 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
2-Bromo-2-phenylacetophenone
40317-0050
40317-0250
TS40317-0050EA
69
USD
TS40317-0050
TS40317-0250
Desyl bromide 97%
Desyl bromide
Formula:
C₁₄H₁₁BrO MW: 275.14 Melting Pt: 53…58 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00000136 CAS Number: 1484-50-0 EINECS: 216-057-1 UN: 1759 ADR: 8 III |
Specification Test Results
Appearance | White to beige to light brown Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 53 to 58 °C |
GC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...