2-(4-Aminophenyl)ethanol 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Aminophenethyl alcohol

40093-0250 40093-1000
TS40093-0250EA 119 USD
TS40093-0250 TS40093-1000
2-(4-Aminophenyl)ethanol 97%
2-(4-Aminophenyl)ethanol
Formula: C₈H₁₁NO
Melting Pt: 104…108 °C
Storage Temperature: Ambient
MDL Number: MFCD00007922
CAS Number: 104-10-9
EINECS: 203-174-8

Order Now

Specification Test Results

Appearance Off-white to brown Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 106 to 110 °C
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR