2'-Aminoacetophenone 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-(2-Aminophenyl)ethanone, o-Aminoacetophenone

40062-0050 40062-0100 40062-0500
TS40062-0050EA 41 USD
TS40062-0050 TS40062-0100 TS40062-0500
2'-Aminoacetophenone 97%
2'-Aminoacetophenone
Formula: C₈H₉NO
MW: 135.17 g/mol
Boiling Pt: 70…71 °C (3 mmHg)
Melting Pt: 20 °C
Flash Pt: 112 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007717
CAS Number: 551-93-9
EINECS: 209-002-8
Merck Index: 13,00412

Order Now

Specification Test Results

Appearance Yellow to yellow-brown Liquid
Infrared spectrum Conforms
Refractive index 1.6130 to 1.6170 (20 °C, 589 nm)
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR