2-Bromo-5-formylpyridine 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 6-Bromonicotinaldehyde, 2-Bromopyridine-5-carbaldehyde, 2-Bromopyridine-5-carboxaldehyde, 6-Bromo-3-pyridinecarboxaldehyde

39985-0010 39985-0050 39985-0250
TS39985-0010EA 70 USD
TS39985-0010 TS39985-0050 TS39985-0250
2-Bromo-5-formylpyridine 95%
2-Bromo-5-formylpyridine
Formula: C₆H₄BrNO
MW: 186.01
Melting Pt: 104…110 °C
Storage Temperature: Ambient
MDL Number: MFCD04115419
CAS Number: 149806-06-4

Order Now

Specification Test Results

Appearance White to brown Crystalline powder
Infrared spectrum Conforms
Melting point 98 to 108 °C
GC ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR