2-Bromo-3-formylpyridine 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-Bromopyridine-3-carboxaldehyde, 2-Bromonicotinaldehyde, 2-Bromo-3-pyridinecarboxaldehyde
39967-0010
39967-0050
TS39967-0010EA
46
USD
TS39967-0010
TS39967-0050
2-Bromo-3-formylpyridine 96%
2-Bromo-3-formylpyridine
Formula:
C₆H₄BrNO MW: 186.01 g/mol Boiling Pt: 100 °C (3 mmHg) Melting Pt: 70…74 °C Storage Temperature: Ambient |
MDL Number:
MFCD04966945 CAS Number: 128071-75-0 |
Specification Test Results
Appearance | White to tan Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 69 to 75 °C |
HPLC | ≥95.0 % |
NMR (1H-NMR) | Consistent with structure |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...