Ethyl (4-bromobenzoyl)acetate 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: ethyl 3-(4-bromophenyl)-3-oxopropanoate, 3-(4-bromophenyl)-3-oxo-propionic acid ethyl ester, Benzenepropanoic acid, 4-bromo-beta-oxo-, ethyl ester

39916-0010 39916-0050
TS39916-0010EA 104 USD
TS39916-0010 TS39916-0050
Ethyl (4-bromobenzoyl)acetate 95%
Ethyl (4-bromobenzoyl)acetate
Formula: C₁₁H₁₁BrO₃
Boiling Pt: 268…269 °C (760 mmHg)
Density: 1.432 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00231243
CAS Number: 26510-95-2

Order Now

Specification Test Results

Appearance Clear yellow Liquid
Infrared spectrum Conforms
HPLC ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR