2-Formylimidazole 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1H-Imidazole-2-carbaldehyde, Imidazole-2-carboxaldehyde

39847-0010 39847-0050
TS39847-0010EA 93 USD
TS39847-0010 TS39847-0050
2-Formylimidazole 98%
2-Formylimidazole
Formula: C₄H₄N₂O
MW: 96.09 g/mol
Boiling Pt: 80 °C (.1 mmHg)
Melting Pt: 209 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00003544
CAS Number: 10111-08-7

Order Now

Specification Test Results

Appearance Off-white to light tan Powder
Infrared spectrum Conforms
HPLC ≥97.5 %
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR