2-Chloro-5-formylpyridine 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
6-Chloronicotinaldehyde, 2-Chloropyridine-5-carboxaldehyde, 6-Chloropyridine-3-carboxaldehyde
39810-0010
39810-0050
TS39810-0010EA
109
USD
TS39810-0010
TS39810-0050
2-Chloro-5-formylpyridine 97%
2-Chloro-5-formylpyridine
Formula:
C₆H₄ClNO MW: 141.56 g/mol Melting Pt: 77…81 °C Storage Temperature: Ambient |
MDL Number:
MFCD03095223 CAS Number: 23100-12-1 |
Specification Test Results
Appearance | White to yellow Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 77 to 82 °C |
GC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...