6-Fluoroveratraldehyde 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Fluoro-4,5-dimethoxybenzaldehyde, 4,5-Dimethoxy-2-fluorobenzaldehyde, 3,4-Dimethoxy-6-fluorobenzaldehyde

39809-0010 39809-0050
TS39809-0010EA 48 USD
TS39809-0010 TS39809-0050
6-Fluoroveratraldehyde 97%
6-Fluoroveratraldehyde
Formula: C₉H₉FO₃
MW: 184.17 g/mol
Melting Pt: 98…100 °C
Storage Temperature: Ambient
MDL Number: MFCD00061108
CAS Number: 71924-62-4

Order Now

Specification Test Results

Appearance White to light yellow Powder or solid
Infrared spectrum Conforms
Melting point 98 to 100 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR